siRNA Details
| Gene Symbol: Pou3f4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008901.1 | NP_032927.1 | 1 | 749 | ||
| GenBank mRNA | AK158302.1 | - | 1 | 782 | ||
| BC052012.1 | - | 1 | 797 | |||
| BC086670.1 | - | 760 | ||||
| BC138657.1 | - | 1 | 824 | 8861677 | ||
| BC145919.1 | - | 1 | 824 | 40131222 | ||
| X66603.1 | - | 792 | ||||

Pou3f4
Genomic Map