siRNA Details
| Gene Symbol: Ppfibp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163557.1 | NP_001157029.1 | 11 | 1144 | ||
| NM_008905.2 | NP_032931.2 | 11 | 1380 | |||
| GenBank mRNA | AK012152.1 | - | 8 | 803 | ||
| BC055935.1 | - | 1 | 14 | |||
| BC100346.1 | - | 11 | 1144 | 30936010 | ||
| U79024.1 | - | 11 | 1165 | |||

Ppfibp2
Genomic Map