siRNA Details
| Gene Symbol: Prkar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253890.1 | NP_001240819.1 | 8 | 859 | ||
| NM_008923.3 | NP_032949.3 | 8 | 861 | |||
| GenBank mRNA | AK161621.1 | - | 8 | 792 | ||
| BC011424.1 | - | 8 | 800 | 3674751 | ||
| CO425808.1 | - | 4 | 274 | |||
| M20473.1 | - | 8 | 807 | |||

Prkar1b
Genomic Map