siRNA Details
| Gene Symbol: Ptdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008959.3 | NP_032985.2 | 13 | 2021 | ||
| GenBank mRNA | AF042731.1 | - | 13 | 1436 | ||
| AK036990.1 | - | 13 | 1483 | |||
| AK054101.1 | - | 13 | 1460 | |||
| AK085857.1 | - | 13 | 2022 | |||
| AK086136.1 | - | 13 | 1680 | |||
| AK088048.1 | - | 13 | 1503 | |||
| AK137681.1 | - | 13 | 1691 | |||
| AK172873.1 | - | 12 | 3807 | |||
| AK219321.1 | - | 2 | 110 | |||
| BC003260.1 | - | 13 | 1658 | 3501141 | ||

Ptdss1
Genomic Map