siRNA Details
| Gene Symbol: Ptdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008959.3 | NP_032985.2 | 13 | 2110 | ||
| GenBank mRNA | AF042731.1 | - | 13 | 1525 | ||
| AK036990.1 | - | 13 | 1572 | |||
| AK054101.1 | - | 13 | 1549 | |||
| AK085857.1 | - | 13 | 2111 | |||
| AK086136.1 | - | 13 | 1769 | |||
| AK088048.1 | - | 13 | 1592 | |||
| AK137681.1 | - | 13 | 1780 | |||
| AK172873.1 | - | 12 | 3896 | |||
| AK219321.1 | - | 2 | 199 | |||
| BC003260.1 | - | 13 | 1747 | 3501141 | ||

Ptdss1
Genomic Map