siRNA Details
| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008960.2 | NP_032986.1 | 5 | 1264 | ||
| GenBank mRNA | AK030750.1 | - | 1 | 27 | ||
| AK076980.1 | - | 5 | 1222 | |||
| AK088717.1 | - | 5 | 1243 | |||
| AK148736.1 | - | 5 | 1099 | |||
| AK164302.1 | - | 5 | 1247 | |||
| BC021445.1 | - | 5 | 613 | 5038272 | ||
| U92437.1 | - | 5 | 1344 | |||

Pten
Genomic Map