siRNA Details
| Gene Symbol: Ptger2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008964.4 | NP_032990.1 | 1 | 1069 | ||
| GenBank mRNA | AK036474.1 | - | 1 | 514 | ||
| AK036531.1 | - | 1 | 988 | |||
| AK054132.1 | - | 1 | 590 | |||
| AK086904.1 | - | 1 | 983 | |||
| AK136577.1 | - | 1 | 82 | |||
| AK137515.1 | - | 1 | 279 | |||
| BC005440.1 | - | 1 | 577 | 3487087 | ||
| D50589.1 | - | 1 | 360 | |||
More Information
Set Membership: |
|

Ptger2
Genomic Map