siRNA Details
| Gene Symbol: Ptp4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164745.1 | NP_001158217.1 | 3, 4 | 888 | ||
| NM_008974.4 | NP_033000.1 | 3, 4 | 919 | |||
| GenBank mRNA | AF035644.1 | - | 2, 3 | 370 | ||
| AK017872.1 | - | 2, 3 | 626 | |||
| AK033092.1 | - | 2, 3 | 624 | |||
| AK045093.1 | - | 2, 3 | 621 | |||
| AK135662.1 | - | 2, 3 | 125 | |||
| AK155895.1 | - | 4, 5 | 962 | |||
| BC086794.1 | - | 2, 3 | 301 | 6811716 | ||
| BC087551.1 | - | 4, 5 | 947 | 6329297 | ||

Ptp4a2
Genomic Map