siRNA Details
Gene Symbol: Ptpn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008976.2 | NP_033002.2 | 8, 9 | 1288 | ||
GenBank mRNA | AF170902.1 | - | 7, 8 | 855 | ||
AF170903.1 | - | 7, 8 | 855 | |||
AK028783.1 | - | 8, 9 | 1252 | |||
AK037646.1 | - | 8, 9 | 1289 | |||
AK086524.1 | - | 8, 9 | 1282 | |||
AK156480.1 | - | 8, 9 | 1034 | |||
D31842.1 | - | 7, 8 | 885 |
