siRNA Details
Gene Symbol: Ptpn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008976.2 | NP_033002.2 | 13 | 1662 | ||
GenBank mRNA | AF170902.1 | - | 12 | 1229 | ||
AF170903.1 | - | 12 | 1229 | |||
AK028783.1 | - | 13 | 1626 | |||
AK037646.1 | - | 13 | 1663 | |||
AK086515.1 | - | 2 | 1430 | |||
AK156480.1 | - | 13 | 1408 | |||
D31842.1 | - | 12 | 1259 |
