siRNA Details
| Gene Symbol: Ptpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M80739.1 | - | 6 | 580 | ||
| M81477.1 | - | 6 | 580 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpn2| Gene Symbol: Ptpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M80739.1 | - | 6 | 580 | ||
| M81477.1 | - | 6 | 580 | |||