siRNA Details
| Gene Symbol: Ptpn20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008978.2 | NP_033004.1 | 4, 5 | 352 | ||
| GenBank mRNA | AK017003.1 | - | 4, 5 | 415 | ||
| AK029493.1 | - | 4, 5 | 350 | |||
| BC139817.1 | - | 2, 3 | 243 | |||
| D64141.1 | - | 3, 4 | 351 | |||

Ptpn20
Genomic Map