siRNA Details
| Gene Symbol: Ptpn20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008978.2 | NP_033004.1 | 9 | 1107 | ||
| GenBank mRNA | AK017003.1 | - | 9 | 1170 | ||
| AK029493.1 | - | 9 | 1105 | |||
| D64141.1 | - | 8 | 1106 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpn20| Gene Symbol: Ptpn20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008978.2 | NP_033004.1 | 9 | 1107 | ||
| GenBank mRNA | AK017003.1 | - | 9 | 1170 | ||
| AK029493.1 | - | 9 | 1105 | |||
| D64141.1 | - | 8 | 1106 | |||