siRNA Details
| Gene Symbol: Pvrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008990.3 | NP_033016.3 | 6 | 1342 | ||
| GenBank mRNA | BC046770.1 | - | 6 | 1238 | 5703547 | |
| BC059941.1 | - | 6 | 1294 | 6530953 | ||
| D26107.1 | - | 6 | 1071 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pvrl2| Gene Symbol: Pvrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008990.3 | NP_033016.3 | 6 | 1342 | ||
| GenBank mRNA | BC046770.1 | - | 6 | 1238 | 5703547 | |
| BC059941.1 | - | 6 | 1294 | 6530953 | ||
| D26107.1 | - | 6 | 1071 | |||