siRNA Details
| Gene Symbol: Rab3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166399.2 | NP_001159871.1 | 5 | 1414 | ||
| NM_009001.6 | NP_033027.1 | 5 | 1409 | |||
| GenBank mRNA | AK005307.1 | - | 5 | 1367 | ||
| AK005362.1 | - | 5 | 1371 | |||
| BC053519.1 | - | 5 | 1373 | 6467763 | ||
| BU843202.1 | - | 246 | ||||

Rab3a
Genomic Map