siRNA Details
| Gene Symbol: EIF1AY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278612.1 | NP_001265541.1 | 3 | 339 | ||
| NM_004681.3 | NP_004672.2 | 3 | 339 | |||
| GenBank mRNA | AF000987.1 | - | 3 | 310 | ||
| BC005248.1 | - | 3 | 221 | 3933718 | ||
| BF694967.1 | - | 325 | ||||
| BG499107.1 | - | 3 | 326 | |||
| BP331805.1 | - | 3 | 339 | |||
| BT007201.1 | - | 3 | 178 | |||
| DQ786267.1 | - | 3 | 262 | |||

EIF1AY
Genomic Map