siRNA Details
| Gene Symbol: Rbbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009030.3 | NP_033056.2 | 3 | 330 | ||
| GenBank mRNA | AK082419.1 | - | 3 | 302 | ||
| AK141176.1 | - | 2 | 784 | |||
| AK166437.1 | - | 3 | 300 | |||
| BC138568.1 | - | 3 | 308 | 8861588 | ||
| BC138570.1 | - | 3 | 309 | 8861590 | ||
| BY277283.1 | - | 3 | 330 | |||
| U35141.1 | - | 3 | 300 | |||

Rbbp4
Genomic Map