siRNA Details
Gene Symbol: Rdh16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009040.3 | NP_033066.1 | 4 | 2341 | ||
GenBank mRNA | AF030513.1 | - | 4 | 2342 | ||
AK149404.1 | - | 3 | 2085 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rdh16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009040.3 | NP_033066.1 | 4 | 2341 | ||
GenBank mRNA | AF030513.1 | - | 4 | 2342 | ||
AK149404.1 | - | 3 | 2085 |