siRNA Details
| Gene Symbol: Rdh16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009040.3 | NP_033066.1 | 4 | 2960 | ||
| GenBank mRNA | AF030513.1 | - | 4 | 2961 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rdh16| Gene Symbol: Rdh16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009040.3 | NP_033066.1 | 4 | 2960 | ||
| GenBank mRNA | AF030513.1 | - | 4 | 2961 | ||