siRNA Details
| Gene Symbol: Slc50a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009057.3 | NP_033083.2 | 3 | 316 | ||
| GenBank mRNA | AK014305.1 | - | 3 | 269 | ||
| AK045924.1 | - | 1 | 629 | |||
| AK079716.1 | - | 2 | 381 | |||
| AK158862.1 | - | 2 | 341 | |||
| AK170162.1 | - | 3 | 267 | |||
| AK177059.1 | - | 3 | 224 | |||
| BC014292.1 | - | 3 | 300 | 4190124 | ||
| BY333933.1 | - | 3 | 187 | |||
| X96618.1 | - | 3 | 307 | |||

Slc50a1
Genomic Map