siRNA Details
| Gene Symbol: Ring1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009066.3 | NP_033092.3 | 5 | 1057 | ||
| GenBank mRNA | AK154962.1 | - | 5 | 1018 | ||
| AK155509.1 | - | 5 | 1033 | |||
| BC009070.1 | - | 5 | 1020 | 2651194 | ||
| BC082771.1 | - | 2 | 1528 | |||
| BU918502.1 | - | 1 | 100 | |||
| CT010219.1 | - | 3 | 747 | |||
| KJ650216.1 | - | 4 | 834 | |||
| Y12881.1 | - | 3 | 753 | |||

Ring1
Genomic Map