siRNA Details
| Gene Symbol: Ripk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009068.3 | NP_033094.3 | 4 | 517 | ||
| GenBank mRNA | AK030959.1 | - | 4 | 748 | ||
| AK048989.1 | - | 4 | 745 | |||
| AK156803.1 | - | 4 | 782 | |||
| BC050905.1 | - | 4 | 517 | 5721177 | ||
| BC054542.1 | - | 4 | 517 | 5721177 | ||
| BC058162.1 | - | 4 | 703 | 5135235 | ||
| U25995.1 | - | 3 | 432 | |||

Ripk1
Genomic Map