siRNA Details
| Gene Symbol: Rom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009073.4 | NP_033099.3 | 3 | 1241 | ||
| GenBank mRNA | AK133529.1 | - | 3 | 1242 | ||
| BC014827.1 | - | 3 | 1063 | 4527997 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rom1| Gene Symbol: Rom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009073.4 | NP_033099.3 | 3 | 1241 | ||
| GenBank mRNA | AK133529.1 | - | 3 | 1242 | ||
| BC014827.1 | - | 3 | 1063 | 4527997 | ||