siRNA Details
| Gene Symbol: Rps17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009092.3 | NP_033118.1 | 5 | 438 | ||
| GenBank mRNA | AK007992.1 | - | 5 | 446 | ||
| AK010647.1 | - | 5 | 440 | |||
| AK012781.1 | - | 5 | 427 | |||
| AK158902.1 | - | 4 | 504 | |||
| AK162022.1 | - | 5 | 432 | |||
| AK162335.1 | - | 5 | 430 | |||
| AK167206.1 | - | 5 | 432 | |||
| BC081466.1 | - | 4 | 425 | 6412006 | ||
| BC086900.1 | - | 5 | 431 | 6760363 | ||
| BC086901.1 | - | 5 | 431 | 6763239 | ||
| D25213.1 | - | 4 | 419 | |||

Rps17
Genomic Map