siRNA Details
| Gene Symbol: Rsu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009105.4 | NP_033131.2 | 4 | 396 | ||
| GenBank mRNA | AK009288.1 | - | 4 | 289 | ||
| AK011857.1 | - | 4 | 289 | |||
| AK135491.1 | - | 5 | 379 | |||
| AK149835.1 | - | 4 | 266 | |||
| AK150011.1 | - | 4 | 264 | |||
| AK152655.1 | - | 4 | 276 | |||
| AK169076.1 | - | 4 | 314 | |||
| BC003827.1 | - | 4 | 259 | 3493122 | ||
| CX228720.1 | - | 4 | 396 | |||
| X63039.1 | - | 4 | 211 | |||

Rsu1
Genomic Map