siRNA Details
| Gene Symbol: Rsu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009105.4 | NP_033131.2 | 6 | 652 | ||
| GenBank mRNA | AK009288.1 | - | 6 | 545 | ||
| AK011857.1 | - | 6 | 545 | |||
| AK013076.1 | - | 2 | 123 | |||
| AK135491.1 | - | 7 | 635 | |||
| AK150011.1 | - | 6 | 520 | |||
| AK152655.1 | - | 6 | 532 | |||
| AK169076.1 | - | 6 | 570 | |||
| BC003827.1 | - | 6 | 515 | 3493122 | ||
| CX228720.1 | - | 6 | 652 | |||
| X63039.1 | - | 6 | 467 | |||

Rsu1
Genomic Map