siRNA Details
| Gene Symbol: S100a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009113.4 | NP_033139.2 | 3, 4 | 497 | ||
| GenBank mRNA | AK002755.1 | - | 2, 3 | 296 | ||
| AK008895.1 | - | 2, 3 | 296 | |||
| BC005687.1 | - | 3, 4 | 497 | 3155915 | ||
| CK334302.1 | - | 2, 3 | 464 | |||
| CX219167.1 | - | 2, 3 | 220 | |||
| X99921.1 | - | 2, 3 | 224 | |||

S100a13
Genomic Map