siRNA Details
| Gene Symbol: Nkx1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009123.2 | NP_033149.1 | 2 | 1031 | ||
| GenBank mRNA | AF222444.1 | - | 2 | 1031 | ||
| BC139757.1 | - | 2 | 960 | 40047346 | ||
| X75384.1 | - | 2 | 1136 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx1-2| Gene Symbol: Nkx1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009123.2 | NP_033149.1 | 2 | 1031 | ||
| GenBank mRNA | AF222444.1 | - | 2 | 1031 | ||
| BC139757.1 | - | 2 | 960 | 40047346 | ||
| X75384.1 | - | 2 | 1136 | |||