siRNA Details
| Gene Symbol: Atxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199304.1 | NP_001186233.1 | 7 | 2995 | ||
| NM_001199305.1 | NP_001186234.1 | 8 | 3144 | |||
| NM_009124.6 | NP_033150.2 | 8 | 3120 | |||
| GenBank mRNA | AK048268.1 | - | 8 | 3107 | ||
| AK169931.1 | - | 8 | 3140 | |||
| BC058178.1 | - | 7 | 2961 | 5326559 | ||
| X83542.1 | - | 7 | 2861 | |||

Atxn1
Genomic Map