siRNA Details
| Gene Symbol: Scg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009129.2 | NP_033155.1 | 2 | 487 | ||
| GenBank mRNA | BC014717.1 | - | 2 | 456 | ||
| BC048249.1 | - | 2 | 387 | 6493827 | ||
| BI735349.1 | - | 2 | 458 | |||
| CK619591.1 | - | 2 | 487 | |||

Scg2
Genomic Map