siRNA Details
| Gene Symbol: Selplg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009151.3 | NP_033177.3 | 2 | 319 | ||
| GenBank mRNA | AK089188.1 | - | 2 | 294 | ||
| AK089214.1 | - | 2 | 245 | |||
| AK155762.1 | - | 2 | 250 | |||
| AK156933.1 | - | 2 | 250 | |||
| AK158134.1 | - | 2 | 250 | |||
| AK171693.1 | - | 2 | 263 | |||
| AK172077.1 | - | 2 | 253 | |||
| BC003874.1 | - | 2 | 251 | 3495181 | ||
| CJ134610.1 | - | 2 | 320 | |||
| X91144.1 | - | 1 | 168 | |||

Selplg
Genomic Map