siRNA Details
| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009164.2 | NP_033190.2 | 12 | 1214 | ||
| GenBank mRNA | AK145156.1 | - | 12 | 1154 | ||
| AK161215.1 | - | 11 | 1138 | |||
| AK166337.1 | - | 12 | 1207 | |||
| AK172060.1 | - | 12 | 1212 | |||
| BC004598.1 | - | 10 | 935 | 3584958 | ||
| X87671.1 | - | 9 | 1073 | |||

Sh3bp1
Genomic Map