siRNA Details
| Gene Symbol: Shfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009169.2 | NP_033195.1 | 3 | 389 | ||
| GenBank mRNA | AK135462.1 | - | 342 | |||
| AK160432.1 | - | 3 | 382 | |||
| AK161690.1 | - | 3 | 362 | |||
| AK167914.1 | - | 3 | 339 | |||
| BC023490.1 | - | 3 | 346 | 5251089 | ||
| BC058117.1 | - | 3 | 355 | 6412144 | ||
| U41626.1 | - | 3 | 331 | |||
| X83589.1 | - | 3 | 364 | |||

Shfm1
Genomic Map