siRNA Details
| Gene Symbol: RARRES3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004585.4 | NP_004576.2 | 4 | 745 | ||
| GenBank mRNA | AB030815.1 | - | 4 | 701 | ||
| AF060228.1 | - | 4 | 706 | |||
| AF092922.1 | - | 4 | 738 | |||
| BC009678.1 | - | 4 | 725 | 3877277 | ||

RARRES3
Genomic Map