siRNA Details
| Gene Symbol: Siah1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009172.2 | NP_033198.1 | 2 | 1854 | ||
| GenBank mRNA | AK019266.1 | - | 1 | 652 | ||
| Z19579.1 | - | 2 | 1782 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Siah1a| Gene Symbol: Siah1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009172.2 | NP_033198.1 | 2 | 1854 | ||
| GenBank mRNA | AK019266.1 | - | 1 | 652 | ||
| Z19579.1 | - | 2 | 1782 | |||