siRNA Details
| Gene Symbol: Siah1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC052887.1 | - | 4 | 1564 | 30053109 | |
| DV648408.1 | - | 1 | 117 | |||
| U50958.1 | - | 1 | 81 | |||
| Z19580.1 | - | 4 | 1569 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Siah1b| Gene Symbol: Siah1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC052887.1 | - | 4 | 1564 | 30053109 | |
| DV648408.1 | - | 1 | 117 | |||
| U50958.1 | - | 1 | 81 | |||
| Z19580.1 | - | 4 | 1569 | |||