siRNA Details
| Gene Symbol: St6galnac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009180.3 | NP_033206.2 | 2 | 270 | ||
| GenBank mRNA | AK004613.1 | - | 2 | 271 | ||
| AK136381.1 | - | 273 | ||||
| BC003276.1 | - | 2 | 239 | |||
| BC010208.1 | - | 222 | ||||
| X93999.1 | - | 2 | 234 | |||

St6galnac2
Genomic Map