siRNA Details
| Gene Symbol: Six1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009189.3 | NP_033215.2 | 2 | 1828 | ||
| GenBank mRNA | AK031122.1 | - | 2 | 1583 | ||
| AK144753.1 | - | 2 | 1839 | |||
| BC023304.1 | - | 2 | 1813 | 4188451 | ||
| X80339.1 | - | 2 | 1511 | |||

Six1
Genomic Map