siRNA Details
| Gene Symbol: Six1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009189.3 | NP_033215.2 | 2 | 2508 | ||
| GenBank mRNA | AK031122.1 | - | 2 | 2263 | ||
| AV495228.1 | - | 1 | 181 | |||
| X80339.1 | - | 2 | 2189 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Six1| Gene Symbol: Six1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009189.3 | NP_033215.2 | 2 | 2508 | ||
| GenBank mRNA | AK031122.1 | - | 2 | 2263 | ||
| AV495228.1 | - | 1 | 181 | |||
| X80339.1 | - | 2 | 2189 | |||