siRNA Details
| Gene Symbol: Slc16a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009197.2 | NP_033223.2 | 6 | 2316 | ||
| GenBank mRNA | AF045692.1 | - | 6 | 2367 | ||
| BC080678.1 | - | 6 | 2152 | 6313495 | ||
| BC096587.1 | - | 6 | 2254 | 3992886 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc16a2| Gene Symbol: Slc16a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009197.2 | NP_033223.2 | 6 | 2316 | ||
| GenBank mRNA | AF045692.1 | - | 6 | 2367 | ||
| BC080678.1 | - | 6 | 2152 | 6313495 | ||
| BC096587.1 | - | 6 | 2254 | 3992886 | ||