siRNA Details
| Gene Symbol: Slc4a1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009206.2 | NP_033232.2 | 5, 6 | 1211 | ||
| GenBank mRNA | AF035526.1 | - | 6, 7 | 1094 | ||
| AK138398.1 | - | 5, 6 | 1166 | |||
| AK147161.1 | - | 5, 6 | 549 | |||
| AK160250.1 | - | 4, 5 | 1086 | |||
| BC072588.1 | - | 5, 6 | 1194 | |||
| BC090964.1 | - | 5, 6 | 763 | 30625511 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106709.

Slc4a1ap
Genomic Map