siRNA Details
| Gene Symbol: TROVE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042369.2 | NP_001035828.1 | 4 | 1188 | ||
| NM_001042370.2 | NP_001035829.2 | 4 | 1188 | |||
| NM_001173524.1 | NP_001166995.1 | 4 | 1194 | |||
| NM_001173525.1 | NP_001166996.1 | 4 | 1188 | |||
| NM_004600.5 | NP_004591.2 | 4 | 1188 | |||
| NR_033393.1 | 3 | 593 | ||||
| XM_006711495.3 | XP_006711558.1 | 5 | 1260 | |||
| XM_006711496.3 | XP_006711559.1 | 6 | 1436 | |||
| XM_006711497.3 | XP_006711560.1 | 5 | 1370 | |||
| XM_011509922.2 | XP_011508224.1 | 4 | 769 | |||
| XM_017002180.1 | XP_016857669.1 | 5 | 1069 | |||
| XM_017002181.1 | XP_016857670.1 | 5 | 1298 | |||
| XM_017002182.1 | XP_016857671.1 | 4 | 1003 | |||
| XM_017002183.1 | XP_016857672.1 | 2 | 340 | |||
| GenBank mRNA | AK314594.1 | - | 4 | 981 | ||
| AY205314.1 | - | 4 | 1002 | |||
| AY205315.1 | - | 4 | 1002 | |||
| BC036658.2 | - | 4 | 1030 | 4821776 | ||
| BX394323.2 | - | 3 | 166 | |||
| CR936683.1 | - | 3 | 562 | |||
| J04137.1 | - | 3 | 857 | |||
| M25077.1 | - | 4 | 1002 | |||

TROVE2
Genomic Map