siRNA Details
| Gene Symbol: Sms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009214.3 | NP_033240.3 | 11 | 1753 | ||
| GenBank mRNA | AK017775.1 | - | 11 | 1724 | ||
| AK049787.1 | - | 11 | 1742 | |||
| AK136798.1 | - | 11 | 1756 | |||
| AK166665.1 | - | 11 | 1779 | |||

Sms
Genomic Map