siRNA Details
| Gene Symbol: Snrpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009227.3 | NP_033253.1 | 4 | 236 | ||
| GenBank mRNA | AK019462.1 | - | 4 | 220 | ||
| BC008262.1 | - | 4 | 212 | 2650413 | ||
| BC051207.1 | - | 4 | 194 | 1396204 | ||
| BC055765.1 | - | 4 | 178 | 6407944 | ||
| BY168551.1 | - | 4 | 236 | |||
| X65704.1 | - | 4 | 140 | |||

Snrpe
Genomic Map