siRNA Details
| Gene Symbol: Spn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037810.2 | NP_001032899.1 | 2 | 793 | ||
| NM_009259.5 | NP_033285.1 | 2 | 793 | |||
| GenBank mRNA | AK041480.1 | - | 2 | 785 | ||
| AK041712.1 | - | 2 | 785 | |||
| AK077764.1 | - | 2 | 784 | |||
| AK134098.1 | - | 2 | 788 | |||
| BC100738.1 | - | 1 | 685 | 40045521 | ||
| BC100739.1 | - | 1 | 685 | 40045520 | ||
| BC100740.1 | - | 1 | 685 | 40045519 | ||
| BC100741.1 | - | 1 | 685 | 40045517 | ||
| S70677.1 | - | 1 | 685 | |||

Spn
Genomic Map