siRNA Details
| Gene Symbol: Sprr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009264.2 | NP_033290.1 | 2 | 520 | ||
| GenBank mRNA | AK144782.1 | - | 2 | 534 | ||
| BC027534.1 | - | 2 | 520 | 1362548 | ||
| X91824.1 | - | 2 | 513 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sprr1a| Gene Symbol: Sprr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009264.2 | NP_033290.1 | 2 | 520 | ||
| GenBank mRNA | AK144782.1 | - | 2 | 534 | ||
| BC027534.1 | - | 2 | 520 | 1362548 | ||
| X91824.1 | - | 2 | 513 | |||