siRNA Details
| Gene Symbol: Sqle | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009270.3 | NP_033296.1 | 11 | 2424 | ||
| GenBank mRNA | AK138457.1 | - | 11 | 2437 | ||
| AK161837.1 | - | 10 | 2314 | |||
| AK163502.1 | - | 11 | 2404 | |||
| BC042781.1 | - | 11 | 1681 | 4915911 | ||
| BC056361.1 | - | 11 | 2424 | 6816800 | ||
| D42048.1 | - | 11 | 2290 | |||

Sqle
Genomic Map