siRNA Details
| Gene Symbol: Sqle | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009270.3 | NP_033296.1 | 11 | 2574 | ||
| GenBank mRNA | AK138457.1 | - | 11 | 2587 | ||
| AK161837.1 | - | 10 | 2464 | |||
| AK163502.1 | - | 11 | 2554 | |||
| AK185651.1 | - | 1 | 91 | |||
| AK190780.1 | - | 1 | 91 | |||
| AK198186.1 | - | 1 | 91 | |||
| AK207067.1 | - | 1 | 91 | |||
| AK207809.1 | - | 1 | 98 | |||
| AK209481.1 | - | 1 | 91 | |||
| AK211636.1 | - | 158 | ||||
| AK212058.1 | - | 1 | 91 | |||
| AK212261.1 | - | 1 | 91 | |||
| AK212508.1 | - | 1 | 91 | |||
| AK213010.1 | - | 1 | 91 | |||
| AK216449.1 | - | 1 | 91 | |||
| AK217980.1 | - | 1 | 92 | |||
| AK219261.1 | - | 1 | 91 | |||
| AK219354.1 | - | 1 | 91 | |||
| BC042781.1 | - | 11 | 1831 | 4915911 | ||
| BC056361.1 | - | 11 | 2574 | 6816800 | ||
| D42048.1 | - | 11 | 2440 | |||

Sqle
Genomic Map