siRNA Details
| Gene Symbol: Srm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009272.4 | NP_033298.1 | 7 | 1055 | ||
| GenBank mRNA | AK028359.1 | - | 7 | 1066 | ||
| AK051101.1 | - | 7 | 1051 | |||
| AK146395.1 | - | 7 | 1012 | |||
| AK219795.1 | - | 1 | 66 | |||
| BC005566.1 | - | 7 | 1059 | 3966710 | ||
| L19311.1 | - | 7 | 1023 | |||

Srm
Genomic Map