siRNA Details
| Gene Symbol: Srp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009273.4 | NP_033299.1 | 2 | 164 | ||
| GenBank mRNA | AK019310.1 | - | 2 | 147 | ||
| AK156845.1 | - | 104 | ||||
| AK167821.1 | - | 2 | 113 | |||
| AK168144.1 | - | 2 | 132 | |||
| BB848168.1 | - | 2 | 181 | |||
| BC021537.1 | - | 2 | 140 | 5353575 | ||
| M29264.1 | - | 2 | 89 | |||

Srp14
Genomic Map